Learn More
Bovine Papilloma Virus Type 1 E2 Monoclonal Antibody (1E4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA171535
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-71535 detects BPV-1 E2 protein in viral/infected samples. MA1-71535 has been successfully used in Western Blot and Immunofluorescence assays. The immunogen used to produce MA1-71535 is from amino acid region 250-280.
Bovine Papilloma Virus Type 1 E2 Monoclonal antibody specifically detects Bovine Papilloma Virus Type 1 E2 in Virus samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Bovine Papilloma Virus Type 1 E2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and no preservative | |
P03122 | |
E2 | |
BPV-1 E2 protein. | |
100 μg | |
Primary | |
Virus | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
1E4 | |
Unconjugated | |
E2 | |
BPV; BPV Protein E2; BPV-1; BPV1-E2 | |
Mouse | |
Protein A | |
RUO | |
1489020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |