Learn More
BPI Monoclonal Antibody (4H5), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA140128
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-40128 detects BPI from human samples.The MA1-40128 immunogen is full length human BPI. MA1-40128 recognizes only free BPI and does not interact with BPI that has formed a complex with LPS. MA1-40128 has been successfully used in ELISA applications. ELISA: monoclonal antibody 4H5 can be used both as coating and as detector.
BPI Monoclonal antibody specifically detects BPI in Human samples. It is validated for ELISA.Spezifikation
BPI | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P17213 | |
BPI | |
Full length human BPI. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
4H5 | |
Unconjugated | |
BPI | |
9230105K17Rik; Bactericidal permeability-increasing protein; bactericidal permeablility increasing protein; bactericidal/permeability-increasing protein; BPI; BPI fold containing family D, member 1; BPIFD1; CAP 57; rBPI; recombinant BPI holoprotein, rBPI; RP13-218F6_A.1 | |
Mouse | |
Protein G | |
RUO | |
671 | |
4° C | |
Liquid |