missing translation for 'onlineSavingsMsg'
Learn More

BRCA1 Monoclonal Antibody (KEN), DyLight™ 650, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA516051

Weitere Details : Netto-Gewicht : 0.10000kg

Artikelnummer. 13287648

  • 473.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Suggested positive control: U2OS and MCF-7 whole cell lysates.

BRCA1 Monoclonal antibody specifically detects BRCA1 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

BRCA1
Monoclonal
1.0 mg/mL
50mM sodium borate with 0.05% sodium azide
P38398
BRCA1
Human BRCA1 (residues 1314-1864).
100 μL
Primary
Human
Antibody
IgG3 κ
Immunocytochemistry, Western Blot
KEN
DyLight 650
BRCA1
brca 1; Brca1; brca-1; BRCA1, DNA repair associated; BRCA1/BRCA2-containing complex, subunit 1; BRCAI; BRCC1; breast and ovarian cancer susceptibility protein 1; breast and ovarian cancer sususceptibility protein 1; breast cancer 1; breast cancer 1 early onset; breast cancer 1, early onset; breast cancer associated 1; breast cancer type 1 susceptibility protein; Breast cancer type 1 susceptibility protein homolog; BROVCA1; BROVCA12; Delta772-3095; early onset breast cancer 1; Fanconi anemia, complementation group S; FANCS; IRIS; OTTHUMP00000212147; OTTHUMP00000212148; OTTHUMP00000212149; OTTHUMP00000212150; OTTHUMP00000212151; OTTHUMP00000212155; PNCA4; PPP1R53; protein phosphatase 1, regulatory subunit 53; PSCP; RING finger protein 53; RING-type E3 ubiquitin transferase BRCA1; RNF53; truncated BRCA1; truncated breast and ovarian cancer susceptibility protein 1; truncated breast and ovarian cancer sususceptibility protein 1; truncated breast cancer type 1 susceptibility protein
Mouse
Protein A
RUO
672
4° C, store in dark
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote