missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA511545
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
STAP1 Polyclonal antibody specifically detects STAP1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
STAP1 | |
Polyclonal | |
Unconjugated | |
STAP1 | |
AI586015; BCR downstream signaling 1; BCR downstream-signaling protein 1; BRDG1; docking protein BRDG1; signal transducing adaptor family member 1; signal-transducing adaptor protein 1; Stap1; STAP-1; Stem cell adaptor protein 1; stem cell adaptor protein STAP-1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
26228, 56792 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.0 mg/mL | |
PBS with 0.09% sodium azide | |
Q9JM90, Q9ULZ2 | |
STAP1 | |
KLH conjugated synthetic peptide between 41-70 amino acids from the N-terminal region of human BRDG 1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |