Learn More
BRDT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540818
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NPPPPEYINT KKNGRLTNQL QYLQKVVLKD LWKHSFSWPF QRPVDAVKLQ Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%
BRDT Polyclonal antibody specifically detects BRDT in Human samples. It is validated for Western Blot.Spezifikation
BRDT | |
Polyclonal | |
Unconjugated | |
BRDT | |
7420412D09Rik; BRD6; Brdt; bromodomain testis associated; bromodomain testis-specific protein; bromodomain, testis-specific; bromodomain-containing female sterile homeotic-like protein; Cancer/testis antigen 9; CT9; female sterile homeotic-related gene 3; fi25h02; Fsrg3; LOW QUALITY PROTEIN: bromodomain testis-specific protein; RING3-like protein; si:dkeyp-85h7.3; testis-specific bromodomain protein; wu:fi25h02; wu:fi25h02 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
676 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q58F21 | |
BRDT | |
Synthetic peptide directed towards the N-terminal of human BRDT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |