Learn More
BRPF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527783
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, BRPF1 tansfected 293T cell. Predicted reactivity: Mouse (97%), Rat (96%), Chicken (89%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BRPF1 Polyclonal antibody specifically detects BRPF1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
BRPF1 | |
Polyclonal | |
Unconjugated | |
BRPF1 | |
4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P55201 | |
BRPF1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 318 of Human BRPF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |