missing translation for 'onlineSavingsMsg'
Learn More

BRUCE Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA534412

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15345341

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

A suggested positive control is human small intestine tissue. PA5-34412 can be used with blocking peptide PEP-1454.

BRUCE Polyclonal antibody specifically detects BRUCE in Human samples. It is validated for Immunohistochemistry.
Spezifikation

Spezifikation

BRUCE
Polyclonal
Unconjugated
BIRC6
A430032G04Rik; A430040A19Rik; AA501170; APOLLON; baculoviral IAP repeat containing 6; baculoviral IAP repeat-containing 6; baculoviral IAP repeat-containing protein 6; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; accccbaculoviral IAP repeat-containing 6; BIR repeat-containing ubiquitin-conjugating enzyme; Birc6; BRUCE; D630005A10Rik; KIAA1289; mKIAA1289; RING-type E3 ubiquitin transferase BIRC6; Ubiquitin-conjugating BIR domain enzyme apollon; ubiquitin-conjugating BIR-domain enzyme apollon
Rabbit
Antigen affinity chromatography
RUO
57448
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry
1 mg/mL
PBS with 0.02% sodium azide
Q9NR09
BIRC6
A 17 amino acid synthetic peptide near the carboxy terminus of human BRUCE
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote