Learn More
BRUNOL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541893
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: PMAAFAAAQM QQMAALNMNG LAAAPMTPTS GGSTPPGITA PAVPSIPSPI Sequence homology: Cow: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
BRUNOL4 Polyclonal antibody specifically detects BRUNOL4 in Human samples. It is validated for Western Blot.Spezifikation
BRUNOL4 | |
Polyclonal | |
Unconjugated | |
CELF4 | |
A230070D14Rik; Brul4; Bruno -like 4; brunol4; BRUNOL-4; bruno-like 4, RNA binding protein; bruno-like protein 4; C130060B05Rik; celf4; CELF-4; CUGBP; CUG-BP and ETR-3 like factor 4; CUG-BP- and ETR-3-like factor 4; CUGBP Elav-like family member 4; CUGBP, Elav-like family member 4; LYST-interacting protein LIP9; RNA-binding protein BRUNOL4; RNA-binding protein BRUNOL-4; zgc:92761 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56853 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BZC1 | |
CELF4 | |
Synthetic peptide directed towards the middle region of human BRUNOL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |