Learn More
C1 inhibitor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531541
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: human plasma, mouse plasma, rat plasma. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1 inhibitor Polyclonal antibody specifically detects C1 inhibitor in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C1 inhibitor | |
Polyclonal | |
Unconjugated | |
SERPING1 | |
C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1Inh; C1-inhibiting factor; C1nh; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12258, 295703, 710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P05155, P97290, Q6P734 | |
SERPING1 | |
Recombinant protein fragment corresponding to a region within amino acids 259 and 481 of Human C1 inhibitor | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |