missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544380
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LCFRPGPALP STRARQSFPG PRQPNGAQDW RRADSLPRPQ GITVQFAGRG Sequence homology: Human: 100%
C10orf47 Polyclonal antibody specifically detects C10orf47 in Human samples. It is validated for Western Blot.Spezifikation
C10orf47 | |
Polyclonal | |
Unconjugated | |
PROSER2 | |
5430407P10Rik; AU067770; C10orf47; proline and serine rich 2; proline and serine-rich protein 2; Proser2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
254427 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86WR7 | |
PROSER2 | |
Synthetic peptide directed towards the C-terminal of human C10orf47 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |