missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543400
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: TWYRSSRGEV QTCSERRPIR NLTFQDLHLH HGGHQAANTS HDLAQRHGLE Sequence homology: Cow: 85%; Dog: 86%; Horse: 92%; Human: 100%
VISTA Polyclonal antibody specifically detects VISTA in Human samples. It is validated for Western Blot.Spezifikation
VISTA | |
Polyclonal | |
Unconjugated | |
VSIR | |
4632428N05Rik; B7H5; B7-H5; C10orf54; C1H10orf54; C28H10orf54; C4H10orf54; chromosome 10 open reading frame 54; DD1alpha; Death Domain1alpha; Dies1; differentiation of ESCs 1; GI24; PD-1H; PDCD1 homolog; PD-H1; platelet receptor Gi24; PP2135; RIKEN cDNA 4632428N05 gene; SISP1; sisp-1; Stress-induced secreted protein-1; UNQ730/PRO1412; V-domain Ig suppressor of T cell activation; VISTA; V-set domain-containing immunoregulatory receptor; V-set immunoregulatory receptor; VSIR; V-type immunoglobulin domain-containing suppressor of T-cell activation | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64115 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H7M9 | |
VSIR | |
Synthetic peptide directed towards the N-terminal of human C10orf54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |