missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532681
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Panda, Bat, Rabbit, Guinea pig (100%).
C14orf1 Polyclonal antibody specifically detects C14orf1 in Guinea Pig, Human, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
C14orf1 | |
Polyclonal | |
Unconjugated | |
ERG28 | |
AD-011; C14orf1; chromosome 14 open reading frame 1; ERG28; HSPC288; NET51; Probable ergosterol biosynthetic protein 28; x0006 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100337837, 100714408, 11161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9UKR5 | |
ERG28 | |
Synthetic 15 amino acid peptide from C-Terminus of human C14orf1. | |
50 μg | |
Primary | |
Guinea Pig, Human, Rabbit | |
Antibody | |
IgG |