Learn More
C14orf130 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542324
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MIQCVVCEDW FHGRHLGAIP PESGDFQEMV CQACMKRCSF LWAYAAQLAV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
C14orf130 Polyclonal antibody specifically detects C14orf130 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C14orf130 | |
Polyclonal | |
Unconjugated | |
UBR7 | |
5730410I19Rik; AA589405; AW557761; C14orf130; C21H14ORF130; fb74a06; hypothetical protein LOC322815; LOW QUALITY PROTEIN: putative E3 ubiquitin-protein ligase UBR7; N-recognin-7; Putative E3 ubiquitin-protein ligase UBR7; RGD1359144; RING-type E3 ubiquitin transferase UBR7; ubiquitin protein ligase E3 component n-recognin 7; ubiquitin protein ligase E3 component n-recognin 7 (putative); Ubr7; wu:fb74a06; wu:fi29b06; zgc:55404 | |
Rabbit | |
Protein A | |
RUO | |
55148 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N806 | |
UBR7 | |
Synthetic peptide directed towards the middle region of human C14orf130 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |