Learn More
HAUS4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528494
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HAUS4 Polyclonal antibody specifically detects HAUS4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HAUS4 | |
Polyclonal | |
Unconjugated | |
Haus4 | |
9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS augmin like complex subunit 4; HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; HAUS4; homologous to Augmin subunits; RGD1305288 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H6D7 | |
Haus4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 214 of Human C14orf94 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |