Learn More
KLHL36 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540938
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: FCCEYLEQEV SEDNYLYLQE LASIYSLKRL DAFIDGFILN HFGTLSFTPD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 83%
KLHL36 Polyclonal antibody specifically detects KLHL36 in Human samples. It is validated for Western Blot.Spezifikation
KLHL36 | |
Polyclonal | |
Unconjugated | |
KLHL36 | |
C16orf44; C18H16orf44; CH73-77K24.1; hypothetical protein LOC553604; kelch like family member 36; kelch-like 36; kelch-like family member 36; kelch-like protein 36; Klhl36; LOW QUALITY PROTEIN: kelch-like protein 36; Unknown (protein for MGC:128624); zgc:110268 | |
Rabbit | |
Protein A | |
RUO | |
79786 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N4N3 | |
KLHL36 | |
Synthetic peptide directed towards the N-terminal of human C16ORF44 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |