missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf49 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531990
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C17orf49 Polyclonal antibody specifically detects C17orf49 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C17orf49 | |
Polyclonal | |
Unconjugated | |
C17orf49 | |
0610010K14Rik; 1110020A23Rik; AL033328; AU045833; BAP18; BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; C17orf49; Chromatin complexes subunit BAP18; chromosome 17 open reading frame 49; HEPIS; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49; MLL1/MLL complex subunit C17orf49 homolog; RIKEN cDNA 0610010K14 gene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124944 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IXM2 | |
C17orf49 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 161 of Human C17orf49 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |