missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Njmu-R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544812
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: SSHYCLYSYR GSRLAQQRGD SEDGSPSGTN AETPSGDDFS LSLADTNLPS Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Horse: 92%; Human: 100%; Rabbit: 86%; Rat: 93%
Njmu-R1 Polyclonal antibody specifically detects Njmu-R1 in Human samples. It is validated for Western Blot.Spezifikation
Njmu-R1 | |
Polyclonal | |
Unconjugated | |
C17orf75 | |
5730455P16Rik; C17orf75; chromosome 17 open reading frame 75; NJMU-R1; Protein Njmu-R1; RGD1310429; RIKEN cDNA 5730455P16 gene; sensitization to ricin complex subunit 2; similar to Protein Njmu-R1; spermatogenesis-related protein; SRI2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64149 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9HAS0 | |
C17orf75 | |
Synthetic peptide directed towards the N-terminal of human C17orf75 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |