Learn More
TPGS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531682
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HCT116. Predicted reactivity: Mouse (85%), Rat (85%), Rhesus Monkey (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TPGS2 Polyclonal antibody specifically detects TPGS2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TPGS2 | |
Polyclonal | |
Unconjugated | |
TPGS2 | |
C18orf10; HMFN0601; HsT3006; L17; PGs2; TPGS2; Tubulin polyglutamylase complex subunit 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q68CL5 | |
TPGS2 | |
Recombinant protein fragment corresponding to a region within amino acids 83 and 284 of Human C18orf10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |