missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C18orf34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531601
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C18orf34 Polyclonal antibody specifically detects C18orf34 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C18orf34 | |
Polyclonal | |
Unconjugated | |
CCDC178 | |
C18orf34; CCDC178; coiled-coil domain containing 178; coiled-coil domain-containing protein 178 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q5BJE1 | |
CCDC178 | |
Recombinant fragment corresponding to a region within amino acids 41 and 254 of Human C18orf34 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |