Learn More
C1D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527595
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Raji. Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (81%), Chicken (84%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1D Polyclonal antibody specifically detects C1D in Human samples. It is validated for Western Blot.Spezifikation
C1D | |
Polyclonal | |
Unconjugated | |
C1D | |
1110036E10Rik; AI875855; C1D; C1D DNA-binding protein; C1D nuclear receptor corepressor; C1D nuclear receptor co-repressor; hC1D; LRP1; mC1D; MGC12261; MGC14659; MGC188504; nuclear DNA binding protein; nuclear DNA binding protein, pseudogene 1; nuclear DNA-binding protein; nuclear nucleic acid-binding protein C1D; rCG23324; RGD1560600; Rrp47; small unique nuclear co-repressor; small unique nuclear receptor corepressor; small unique nuclear receptor co-repressor; SUNCOR; Sun-CoR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10438 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13901 | |
C1D | |
Recombinant fragment corresponding to a region within amino acids 1 and 141 of Human C1D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |