missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1D Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519053
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
C1D Polyclonal antibody specifically detects C1D in Human samples. It is validated for Western Blot.Spezifikation
C1D | |
Polyclonal | |
Unconjugated | |
C1D | |
1110036E10Rik; AI875855; C1D; C1D DNA-binding protein; C1D nuclear receptor corepressor; C1D nuclear receptor co-repressor; hC1D; LRP1; mC1D; MGC12261; MGC14659; MGC188504; nuclear DNA binding protein; nuclear DNA binding protein, pseudogene 1; nuclear DNA-binding protein; nuclear nucleic acid-binding protein C1D; rCG23324; RGD1560600; Rrp47; small unique nuclear co-repressor; small unique nuclear receptor corepressor; small unique nuclear receptor co-repressor; SUNCOR; Sun-CoR | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10438 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q13901 | |
C1D | |
Synthetic peptide sequence (SKNASKVANKGKSKS) corresponding to the C-terminus amino acids of C1D | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |