missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1GALT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535246
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
C1GALT1 Polyclonal antibody specifically detects C1GALT1 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
C1GALT1 | |
Polyclonal | |
Unconjugated | |
C1GALT1 | |
2210410E06Rik; AV284120; B3Gal-T8; Beta-1,3-galactosyltransferase; C1GALT; C1GALT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T; core 1 beta3-Gal-T1; core 1 O-glycan T-synthase; Core 1 synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase (C1galt1); core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; core1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 13-galactosyltransferase (C1galt1); glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1; Plt1; T-syn; T-synthase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
56913, 94192 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q9JJ06, Q9NS00 | |
C1GALT1 | |
KLH conjugated synthetic peptide between 115-144 amino acids from the central region of human C1GALT1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |