missing translation for 'onlineSavingsMsg'
Learn More

C1q Monoclonal Antibody (JL-1), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA140311

Weitere Details : Netto-Gewicht : 0.10000kg

Artikelnummer. 10578294

  • 750.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA1-40311 cross reacts with human, mouse, and rat C1q. MA1-40311 has been successfully used in immunohistochemistry (frozen), ELISA, Immunofluorescence, functional assays and Western blot applications. The MA1-40311 immunogen is purified mouse C1q. Endotoxin <24 EU/mg.

C1q Monoclonal antibody specifically detects C1q in Human, Mouse, Rat samples. It is validated for ELISA, Functional Assay, Immunohistochemistry (Frozen), Western Blot.
Spezifikation

Spezifikation

C1q
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and no preservative
P02745, P02746, P02747, P14106, P31720, P31721, P31722, P98086, Q02105
C1QA, C1QB, C1qc
Purified mouse C1q
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
ELISA, Functional Assay, Immunohistochemistry (Frozen), Western Blot
JL-1
Unconjugated
C1qc
AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B
Mouse
Protein G
RUO
12259, 12260, 12262, 29687, 298566, 362634, 712, 713, 714
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote