missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1q Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA129720
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For tissue staining, antigen retrieval is recommended using Proteinase K for 30 min at 37oC.
C1q Polyclonal antibody specifically detects C1q in Hamster, Human samples. It is validated for Functional Assay, Immunohistochemistry (Paraffin).Spezifikation
C1q | |
Polyclonal | |
whole serum with 0.05% sodium azide | |
P02745, P02746, P02747 | |
C1QA, C1QB, C1qc | |
C1q isolated from the normal human serum. | |
1 mL | |
Primary | |
Hamster, Human | |
Antibody | |
IgG |
Functional Assay, Immunohistochemistry (Paraffin) | |
FITC | |
C1qc | |
AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B | |
Rabbit | |
Ig fraction | |
RUO | |
100766409, 100766700, 100769576, 712, 713, 714 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |