Learn More
C1r Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528028
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Molt-4, Raji. Predicted reactivity: Rat (80%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C1r Polyclonal antibody specifically detects C1r in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
C1r | |
Polyclonal | |
Unconjugated | |
C1R | |
AI132558; C1r; C1ra; complement C1r; complement C1r subcomponent; Complement C1r subcomponent heavy chain; Complement C1r subcomponent light chain; Complement C1r-A subcomponent; Complement C1r-A subcomponent heavy chain; Complement C1r-A subcomponent light chain; Complement component 1 subcomponent r; Complement component 1 subcomponent r-A; complement component 1, r subcomponent; complement component 1, r subcomponent A; complement component 1, r-A subcomponent; mC1rA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
312705, 50909, 715 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P00736, Q8CG16 | |
C1R, C1ra | |
Recombinant fragment corresponding to a region within amino acids 277 and 705 of Human C1r | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |