Learn More
KNP-I Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542778
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: SRGGAEVQIF APDVPQMHVI DHTKGQPSEG ESRNVLTESA RIARGKITDL Sequence homology: Cow: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Rat: 100%; Sheep: 93%; Zebrafish: 86%
KNP-I Polyclonal antibody specifically detects KNP-I in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
KNP-I | |
Polyclonal | |
Unconjugated | |
GATD3A | |
C21orf33; chromosome 21 open reading frame 33; ES1; ES1 protein homolog, mitochondrial; GATD3A; GATD3B; Glutamine amidotransferase-like class 1 domain-containing protein 3A, mitochondrial; Glutamine amidotransferase-like class 1 domain-containing protein 3B, mitochondrial; GT335; H KNPH; HES1; human HES1 protein, homolog to E.coli and zebrafish ES1 protein; Keio novel protein I; Keio novel protein-I; KNPH; KNPI; KNP-I; Protein GT335; Protein HES1; testis secretory sperm-binding protein Li 237E | |
Rabbit | |
Protein A | |
RUO | |
8209 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
GATD3A | |
Synthetic peptide directed towards the N-terminal of human C21orf33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |