Learn More
C21orf59 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542790
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VLLHVKRGDE SQFLLQAPGS TELEELTVQV ARVYNGRLKV QRLCSEMEEL Sequence homology: Cow: 93%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
C21orf59 Polyclonal antibody specifically detects C21orf59 in Human samples. It is validated for Western Blot.Spezifikation
C21orf59 | |
Polyclonal | |
Unconjugated | |
C21ORF59 | |
C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56683 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P57076 | |
C21ORF59 | |
Synthetic peptide directed towards the N-terminal of human C21orf59 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |