Learn More
C22orf13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546258
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: RHRFCTQTLG VDKGYKNQSF YRKHFDTEET RVNQLFAQAK ACKVLVEKCT Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
C22orf13 Polyclonal antibody specifically detects C22orf13 in Human samples. It is validated for Western Blot.Spezifikation
C22orf13 | |
Polyclonal | |
Unconjugated | |
GUCD1 | |
1110038D17Rik; C17H22orf13; C22orf13; C26H22orf13; CG13760 gene product [Drosophila melanogaster] homolog; guanylyl cyclase domain containing 1; guanylyl cyclase domain-containing protein 1; GUCD1; LLN4; protein GUCD1; protein GUCD1 pseudogene; Protein LLN4; uncharacterized protein C22orf13 homolog; wu:fi27d12; zgc:152874 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83606 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96NT3 | |
GUCD1 | |
Synthetic peptide directed towards the middle region of human C22orf13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |