Learn More
C2orf18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543026
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: GLFGFVILSL LLVPMYYIPA GSFSGNPRGT LEDALDAFCQ VGQQPLIAVA Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 85%
C2orf18 Polyclonal antibody specifically detects C2orf18 in Human samples. It is validated for Western Blot.Spezifikation
C2orf18 | |
Polyclonal | |
Unconjugated | |
SLC35F6 | |
4930471M23Rik; 5730478O19Rik; ANT2-binding protein; ANT2BP; C11H2orf18; C17H2orf18; C2orf18; CB018; P40; RGD1309228; sb:cb433; SLC35F6; Solute carrier family 35 member F6; solute carrier family 35, member F6; TANGO9; transmembrane protein C2orf18; transmembrane protein C2orf18 homolog; transport and Golgi organization 9 homolog; UNQ3047/PRO9863; wu:fd19b11; wu:fi67c08; zgc:92765 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54978 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N357 | |
SLC35F6 | |
Synthetic peptide directed towards the middle region of human C2orf18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |