missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA130601
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-30601 detects C3a from mouse samples.
Complement C3a Polyclonal antibody specifically detects Complement C3a in Mouse samples. It is validated for ELISA, Western Blot.Spezifikation
Complement C3a | |
Polyclonal | |
Unconjugated | |
C3 | |
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3r1; Complement C3; complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3 alpha-chain; complement component C3a; complement component C3b; CPAMD1; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; Plp; prepro-C3 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
12266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P01027 | |
C3 | |
Mouse C3 alpha peptide aa 678-692 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgY |