missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3a Receptor Monoclonal Antibody (74), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517476
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17476 targets C3a Receptor in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17476 immunogen is rBL-2H3 transfectants expressing rat C3aR.
C3a Receptor Monoclonal antibody specifically detects C3a Receptor in Rat samples. It is validated for Flow Cytometry.Spezifikation
C3a Receptor | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
O55197 | |
C3ar1 | |
RBL-2H3 transfectants expressing rat C3aR | |
50 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
74 | |
PE | |
C3ar1 | |
anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
Mouse | |
Protein G | |
RUO | |
84007 | |
4° C, do not freeze | |
Liquid |