missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4BPB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528017
Additional Details : Netto-Gewicht : 0.01000kg
Description
Recommended positive controls: Jurkat, Raji, K562, BCL-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C4BPB Polyclonal antibody specifically detects C4BPB in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
C4BPB | |
Polyclonal | |
Unconjugated | |
C4BPB | |
C4b-binding protein beta chain; C4BP; C4BPB; C4bp-ps1; complement component 4 binding protein beta; complement component 4 binding protein, beta; complement component 4 binding protein, pseudogene 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12270, 725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P20851 | |
C4BPB, C4bp-ps1 | |
Recombinant fragment corresponding to a region within amino acids 18 and 252 of Human C4 binding protein B | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |