missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546342
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MARRHCFSYW LLVCWLVVTV AEGQEEVFTP PGDSQNNADA TDCQIFTLTP Sequence homology: Human: 93%
C4orf26 Polyclonal antibody specifically detects C4orf26 in Human samples. It is validated for Western Blot.Spezifikation
C4orf26 | |
Polyclonal | |
Unconjugated | |
ODAPH | |
AI2A4; C4orf26; chromosome 4 open reading frame 26; ODAPH; Odontogenesis associated phosphoprotein; uncharacterized protein C4orf26 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
152816 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q17RF5 | |
ODAPH | |
Synthetic peptide directed towards the N-terminal of human C4orf26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |