missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525710
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
C4orf31 Polyclonal antibody specifically detects C4orf31 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C4orf31 | |
Polyclonal | |
Unconjugated | |
NDNF | |
C4orf31; fibronectin type-III domain-containing protein C4orf31; NDNF; neuron derived neurotrophic factor; neuron-derived neurotrophic factor; NORD; Protein NDNF; UNQ2748/PRO6487 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
79625 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8TB73 | |
NDNF | |
KLH conjugated synthetic peptide between 80-109 amino acids from the N-terminal region of human C4orf31 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |