missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C5 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA533134
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
C5 is part of the membrane attack complex (MAC) of the complement system. C5 is split into C5a og C5b by the C5 convertases from the classical and the alternative complement pathway and the C5b fragment initiates the assembly of the terminal MAC (C5b-C9). The molecular mass of C5 is 190 kDa.Spezifikation
Complement C5 | |
Polyclonal | |
C5 | |
CPAMD4,C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; anaphylatoxin C5a analog; complement C5 | |
Sheep | |
Ion-exchange chromatography | |
RUO | |
727 | |
4°C or -20°C if preferred | |
Liquid |
Immunodiffusion | |
Unconjugated | |
P01031 | |
C5 | |
Human C5, purified from plasma | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |