Learn More
Complement C5a Monoclonal Antibody (295108), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA523910
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In Western blots, this antibody does not cross-react with recombinant mouse Complement Component C3d, recombinant human (rh) Complement Component C3a, or rhComplement Component C5a. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Complement C5a Monoclonal antibody specifically detects Complement C5a in Mouse samples. It is validated for ELISA, Neutralization, Western Blot.Spezifikation
Complement C5a | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P06684 | |
Hc | |
E. coli-derived recombinant mouse Complement Component C5a Asn679-Arg755 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Neutralization, Western Blot | |
295108 | |
Unconjugated | |
Hc | |
anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905 | |
Rat | |
Protein A/G | |
RUO | |
15139 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |