missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C5a Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535000
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-35000 targets C5a Complement in ELISA applications and shows reactivity with Human samples. The PA5-35000 immunogen is human C5a antigen.
Complement C5a Polyclonal antibody specifically detects Complement C5a in Human samples. It is validated for ELISA.Spezifikation
Complement C5a | |
Polyclonal | |
Unconjugated | |
C5 | |
anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905 | |
Rabbit | |
Affinity chromatography | |
RUO | |
727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P01031 | |
C5 | |
human C5a antigen | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG |