Learn More
C5AR1 Monoclonal Antibody (8D6), FITC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517741
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17741 targets C5a receptor/CD88 in FACS applications and shows reactivity with Human samples. The MA5-17741 immunogen is rBL-2H3 (rat basophilic leukemia) transfected cells expressing human C5aR on the cell surface.
C5AR1 Monoclonal antibody specifically detects C5AR1 in Human samples. It is validated for Flow Cytometry.Spezifikation
C5AR1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P21730 | |
C5AR1 | |
RBL-2H3 (rat basophilic leukemia) transfected cells expressing human C5aR on the cell surface | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry | |
8D6 | |
FITC | |
C5AR1 | |
anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 | |
Rat | |
Protein G | |
RUO | |
728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |