Learn More
C5AR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532683
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Monkey (100%); Gibbon (89%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
C5AR1 Polyclonal antibody specifically detects C5AR1 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
C5AR1 | |
Polyclonal | |
Unconjugated | |
C5AR1 | |
anaphylatoxin C5a receptor; C5A; C5a anaphylatoxin chemotactic receptor; C5a anaphylatoxin chemotactic receptor 1; C5a anaphylatoxin receptor; C5a ligand; C5a Receptor; C5AR; C5a-R; C5AR1; C5r1; Cd88; CD88 antigen; complement C5a receptor; complement C5a receptor 1; complement component 5 receptor 1; complement component 5, receptor 1; complement component 5a receptor 1; D7Msu1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P21730 | |
C5AR1 | |
Synthetic 18 amino acid peptide from internal region of human C5AR1 / CD88. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |