missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR77 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533373
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (95%) Monkey, Marmoset (84%).
GPR77 Polyclonal antibody specifically detects GPR77 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
GPR77 | |
Polyclonal | |
Unconjugated | |
C5AR2 | |
C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; C5AR2; C5L2; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; GPF77; GPR77; G-protein coupled receptor 77 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9P296 | |
C5AR2 | |
Synthetic 19 amino acid peptide from C-terminus of human C5L2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |