missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NREP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539301
Weitere Details : Netto-Gewicht : 0.00900kg
Spezifikation
NREP | |
Polyclonal | |
Unconjugated | |
NREP | |
D0H4S114, P311 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27528, 338475, 9315 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q16612, Q07475, Q80Z34 | |
NREP | |
A synthetic peptide derived from the internal region of human C5orf13 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |