missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin 2 Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544166
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EQHFLGCVKR AWDSAEVAPE PQPPPIVSSE DRGPWPLPLY PVLGEYSLDS Sequence homology: Human: 100%
Annexin 2 Receptor Polyclonal antibody specifically detects Annexin 2 Receptor in Human samples. It is validated for Western Blot.Spezifikation
Annexin 2 Receptor | |
Polyclonal | |
Unconjugated | |
ANXA2R | |
annexin A2 receptor; annexin II receptor; Annexin-2 receptor; ANXA2R; AX2R; AXIIR; C5orf39 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
389289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3ZCQ2 | |
ANXA2R | |
Synthetic peptide directed towards the N-terminal of human C5orf39 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |