missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5orf54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532073
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: H1299, MCF-7. Predicted reactivity: Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C5orf54 Polyclonal antibody specifically detects C5orf54 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C5orf54 | |
Polyclonal | |
Unconjugated | |
ZBED8 | |
Buster3; C5orf54; Protein ZBED8; transposon-derived Buster3 transposase-like protein; ZBED8; Zinc finger BED domain-containing protein 8; zinc finger BED-type containing 8; zinc finger, BED-type containing 8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IZ13 | |
ZBED8 | |
Recombinant fragment corresponding to a region within amino acids 394 and 594 of Human C5orf54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |