missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546354
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: SCSLTWETPR WYMAGRVATS TSGCHCWMSR RDLTPLPHPS EPGVLDCLGP Sequence homology: Human: 100%; Mouse: 85%
C6orf1 Polyclonal antibody specifically detects C6orf1 in Human samples. It is validated for Western Blot.Spezifikation
C6orf1 | |
Polyclonal | |
Unconjugated | |
SMIM29 | |
C6orf1; chromosome 6 open reading frame 1; LBH; Protein LBH; Small integral membrane protein 29; SMIM29; uncharacterized protein C6orf1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
221491 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86T20 | |
SMIM29 | |
Synthetic peptide directed towards the N-terminal of human C6orf1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |