Learn More
C8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531290
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Rat (80%), Pig (84%), Rabbit (86%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C8B Polyclonal antibody specifically detects C8B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C8B | |
Polyclonal | |
Unconjugated | |
C8B | |
4930439B20Rik; AI595927; C82; C8b; complement C8 beta chain; complement component 8 subunit beta; complement component 8, beta polypeptide; complement component 8, beta subunit; complement component C8 beta chain; complement component C8 beta subunit; MGC137114 protein; MGC163447 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P07358 | |
C8B | |
Recombinant fragment corresponding to a region within amino acids 362 and 591 of Human C8B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |