missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8orf48 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531610
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C8orf48 Polyclonal antibody specifically detects C8orf48 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
C8orf48 | |
Polyclonal | |
Unconjugated | |
C8orf48 | |
C8orf48; chromosome 8 open reading frame 48; uncharacterized protein C8orf48 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96LL4 | |
C8orf48 | |
Recombinant protein fragment corresponding to a region within amino acids 69 and 275 of Human C8orf48 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |