missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C8orf55 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532684
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Marmoset, Hamster, Panda (88%).
C8orf55 Polyclonal antibody specifically detects C8orf55 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
C8orf55 | |
Polyclonal | |
Unconjugated | |
THEM6 | |
4930572J05Rik; C13H8orf55; C14H8orf55; C8orf55; DSCD75; mesenchymal stem cell protein DSCD75; mesenchymal stem cell protein DSCD75 homolog; protein THEM6; PSEC0098; RGD1359378; Them6; Thioesterase superfamily member 6; UPF0670 protein C8orf55; UPF0670 protein C8orf55 homolog; UPF0670 protein THEM6; UPF0670 protein THEM6 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8WUY1 | |
THEM6 | |
Synthetic 17 amino acid peptide from N-terminus of human C8orf55. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |