missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529066
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Jurkat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Complement C9 Polyclonal antibody specifically detects Complement C9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Complement C9 | |
Polyclonal | |
Unconjugated | |
C9 | |
ARMD15; C9; C9D; complement C9; complement component 9; complement component C9; Complement component C9a; Complement component C9b; complement protein C9; complement protein C9 precursor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P02748 | |
C9 | |
Recombinant fragment corresponding to a region within amino acids 153 and 429 of Human C9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |