missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf114 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531693
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: IMR32, U87-MG. Predicted reactivity: Mouse (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C9orf114 Polyclonal antibody specifically detects C9orf114 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C9orf114 | |
Polyclonal | |
Unconjugated | |
SPOUT1 | |
9930028P05; AL033358; C9orf114; CENP-32; Centromere protein 32; D2Wsu81e; DNA segment, Chr 2, Wayne State University 81, expressed; HSPC109; Kinetochore-associated protein; LOC499770; Putative methyltransferase C9orf114; putative methyltransferase C9orf114 homolog; similar to LOC495800 protein; SPOUT domain containing methyltransferase 1; SPOUT domain-containing methyltransferase 1; Spout1; uncharacterized protein C9orf114 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
227695, 51490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q3UHX9, Q5T280 | |
SPOUT1 | |
Recombinant fragment corresponding to a region within amino acids 113 and 370 of Human C9orf114 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |