missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531617
Additional Details : Netto-Gewicht : 0.01000kg
Description
Recommended positive controls: A549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C9orf43 Polyclonal antibody specifically detects C9orf43 in Human samples. It is validated for Western Blot.Specifications
C9orf43 | |
Polyclonal | |
Unconjugated | |
C9orf43 | |
C9orf43; chromosome 9 open reading frame 43; uncharacterized protein C9orf43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
257169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TAL5 | |
C9orf43 | |
Recombinant fragment corresponding to a region within amino acids 89 and 381 of Human C9orf43 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |